| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
TC08000700.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
ENST00000247781.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
ENST00000297848.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
ENST00000309791.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
ENST00000432943.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
ENST00000537875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08009462.hg.1 |
NM_021110.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000297848.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000309791.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000432943.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000434620.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000498051.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000523142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000537875.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
NM_021110.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000297848.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000309791.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000432943.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000434620.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000498051.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000523142.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
ENST00000537875.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
NM_021110.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800159470.hg.1 |
TC0800008679.hg.1 |
Clariom D TC |