| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019950.hg.1 |
TC06001675.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019950.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019950.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180008.hg.1 |
TC0600011661.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |