Assay Details
| Gene Symbol: TBC1D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143964.3 | NP_001137436.1 | 3 - 4 | 466 | 94 | |
| NM_001143965.3 | NP_001137437.1 | 3 - 4 | 326 | 94 | ||
| NM_001258457.2 | NP_001245386.1 | 3 - 4 | 326 | 94 | ||
| NM_001318805.1 | NP_001305734.1 | 3 - 4 | 383 | 94 | ||
| NM_016495.5 | NP_057579.1 | 3 - 4 | 486 | 94 | ||
| GenBank mRNA | AB449888.1 | - | 2 - 3 | 190 | 94 | |
| AF151073.1 | - | 3 - 4 | 283 | 94 | ||
| AY542308.1 | - | 3 - 4 | 248 | 94 | ||
| BC007054.1 | - | 3 - 4 | 287 | 3828592 | 94 | |
| BG504676.1 | - | 3 - 4 | 499 | 94 | ||
| BI914228.1 | - | 3 - 4 | 447 | 94 | ||
| BP319580.1 | - | 3 - 4 | 257 | 94 | ||
| BX354770.2 | - | 3 - 4 | 323 | 94 | ||
| Gene Symbol: TBC1D7-LOC100130357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318809.1 | NP_001305738.1 | 3 - 4 | 326 | 94 | |
| NR_134872.1 | 3 - 4 | 326 | 94 | |||
| GenBank mRNA | AK057228.1 | - | 3 - 4 | 287 | 93 | |
| AK296391.1 | - | 3 - 4 | 287 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK057228.1 transcripts.
Set Membership: |
|

TBC1D7
Genomic Map