Assay Details
| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094753.1 | - | 1 - 2 | 84 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000237858.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000379979.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000505427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000507605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000508780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | ENST00000512469.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001118890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243658.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_001243659.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011497.hg.1 | NM_002064.2 | Refseq |

GLRX
Genomic Map