Assay Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022158.3 | NP_071441.1 | 1 - 2 | 202 | 135 | |
| GenBank mRNA | AJ404615.1 | - | 1 - 2 | 162 | 135 | |
| AK294871.1 | - | 1 - 2 | 163 | 135 | ||
| BC042680.1 | - | 1 - 2 | 146 | 5195472 | 135 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000957.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17012617.hg.1 | ENST00000300784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012617.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012617.hg.1 | TC17000957.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006904.hg.1 | ENST00000456260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012619.hg.1 | ENST00000300784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012619.hg.1 | ENST00000457624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012619.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012619.hg.1 | TC17000957.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012622.hg.1 | ENST00000300784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012622.hg.1 | ENST00000457624.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012622.hg.1 | ENST00000536165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012622.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17012622.hg.1 | TC17000957.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700170530.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170530.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170530.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170530.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170530.hg.1 | TC1700009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000572813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009275.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170534.hg.1 | TC1700009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000570734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000573841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170540.hg.1 | TC1700009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC2200006904.hg.1 | ENST00000456260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | ENST00000300784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | ENST00000574496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | NM_022158.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700170533.hg.1 | TC1700009275.hg.1 | Clariom D TC |

FN3K
Genomic Map