Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
TC09000357.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000277082.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000376597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000376598.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000415085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000415759.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
ENST00000424347.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
NM_001098802.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003869.hg.1 |
NM_032171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000277082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000376597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000376598.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000415759.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000424347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
NM_001098802.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
NM_032171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000277082.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000376597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000376598.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000415759.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
ENST00000424347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
NM_001098802.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
NM_032171.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151796.hg.1 |
TC0900007663.hg.1 |
Clariom D TC |