| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019095.hg.1 |
ENST00000333461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019095.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019095.hg.1 |
TC17001447.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019094.hg.1 |
ENST00000333461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019094.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019094.hg.1 |
TC17001447.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019096.hg.1 |
ENST00000333461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019096.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019096.hg.1 |
TC17001447.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019092.hg.1 |
ENST00000333461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019092.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019092.hg.1 |
TC17001447.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019093.hg.1 |
ENST00000333461.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019093.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17019093.hg.1 |
TC17001447.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183254.hg.1 |
TC1700010564.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700183259.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183259.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183259.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183259.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183259.hg.1 |
TC1700010564.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
ENST00000584501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010564.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183255.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183255.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183255.hg.1 |
ENST00000333461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700183255.hg.1 |
NM_198993.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700183255.hg.1 |
TC1700010564.hg.1 |
Clariom D TC |