Assay Details
| Gene Symbol: MTMR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007034.1 | XP_016862523.1 | 2 - 3 | 486 | 72 | |
| XM_017007035.1 | XP_016862524.1 | 2 - 3 | 486 | 72 | ||
| XM_017007036.1 | XP_016862525.1 | 2 - 3 | 485 | 72 | ||
| XM_017007037.1 | XP_016862526.1 | 2 - 3 | 486 | 72 | ||
| XM_017007039.1 | XP_016862528.1 | 2 - 3 | 485 | 72 | ||
| XM_017007040.1 | XP_016862529.1 | 2 - 3 | 486 | 72 | ||
More Information
Set Membership: |
|

MTMR14
Genomic Map