| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008372.hg.1 |
TC09000691.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008372.hg.1 |
ENST00000393533.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008372.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008371.hg.1 |
TC09000691.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008371.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159548.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159546.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900159528.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159528.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159528.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
ENST00000393533.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
ENST00000497812.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
ENST00000393533.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
ENST00000497812.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
NM_153436.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159531.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000421776.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000432065.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000434106.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000446274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
ENST00000546203.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
NM_153433.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
NM_153440.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159547.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |