Assay Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC073864.1 | - | 1 - 2 | 119 | 127 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000312916.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000502408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000503538.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | ENST00000506806.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007841.hg.1 | NM_018046.4 | Refseq |

AGGF1
Genomic Map