Assay Details
| Gene Symbol: LRRC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153353.4 | NP_699184.2 | 6 - 7 | 893 | 88 | |
| GenBank mRNA | BC032841.2 | - | 6 - 7 | 632 | 4826965 | 88 |
More Information
Set Membership: |
|

LRRC34
Genomic Map