Assay Details
| Gene Symbol: SLC33A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190992.1 | NP_001177921.1 | 2 - 3 | 1427 | 82 | |
| NM_004733.3 | NP_004724.1 | 2 - 3 | 1427 | 82 | ||
| XM_017007463.1 | XP_016862952.1 | 2 - 3 | 1510 | 82 | ||
| GenBank mRNA | AK312268.1 | - | 2 - 3 | 1397 | 82 | |
| BC014416.1 | - | 2 - 3 | 1374 | 4559958 | 82 | |
| D88152.1 | - | 2 - 3 | 1352 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001929.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034581.hg.1 | TC03001929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034581.hg.1 | ENST00000359479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034581.hg.1 | ENST00000392845.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034581.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03034581.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000460729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | ENST00000496772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012881.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000359479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000392845.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000468581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | ENST00000475842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | NM_001190992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | NM_004733.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196305.hg.1 | TC0300012881.hg.1 | Clariom D TC |

SLC33A1
Genomic Map