Assay Details
| Gene Symbol: KCNAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003471.3 | NP_003462.2 | 2 - 3 | 505 | 108 | |
| GenBank mRNA | AK292999.1 | - | 2 - 3 | 437 | 108 | |
| DA408045.1 | - | 2 - 3 | 483 | 108 | ||
| L39833.1 | - | 2 - 3 | 376 | 108 | ||
| U16953.1 | - | 2 - 3 | 355 | 108 | ||
| U17968.1 | - | 2 - 3 | 280 | 108 | ||
More Information
Set Membership: |
|

KCNAB1
Genomic Map