Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130064.1 | NP_001123536.1 | 2 - 3 | 806 | 128 | |
XM_017006128.1 | XP_016861617.1 | 1 - 2 | 2055 | 128 | ||
GenBank mRNA | AK289699.1 | - | 2 - 3 | 600 | 128 | |
AK290100.1 | - | 2 - 3 | 479 | 128 | ||
BX648458.1 | - | 2 - 3 | 478 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000595.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein kinase C-activating G-protein coupled receptor signaling pathway
response to wounding
glial cell differentiation
axon choice point recognition
regulation of growth
tissue regeneration
cell fate commitment
regulation of filopodium assembly
axon development
phosphatidylserine binding
protein binding
calmodulin binding
lysophosphatidic acid binding
phosphatidylinositol phosphate binding
protein kinase C-activating G-protein coupled receptor signaling pathway
response to wounding
glial cell differentiation
axon choice point recognition
regulation of growth
tissue regeneration
cell fate commitment
regulation of filopodium assembly
axon development
phosphatidylserine binding
protein binding
calmodulin binding
lysophosphatidic acid binding
phosphatidylinositol phosphate binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03010843.hg.1 | TC03000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03010843.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03010843.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03010844.hg.1 | TC03000595.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03010844.hg.1 | ENST00000305124.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03010844.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03010844.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03010844.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160057.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160057.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160057.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160057.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160057.hg.1 | TC0300008412.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300160056.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160056.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160056.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160056.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160056.hg.1 | TC0300008412.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008412.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | ENST00000305124.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | ENST00000393780.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | NM_001130064.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | NM_002045.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300160058.hg.1 | TC0300008412.hg.1 | Clariom D TC |
