Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300764.1 | NP_001287693.1 | 3 - 4 | 450 | 121 | |
| NM_001300766.1 | NP_001287695.1 | 3 - 4 | 450 | 121 | ||
| NM_052965.3 | NP_443197.1 | 3 - 4 | 450 | 121 | ||
| XM_006711148.1 | XP_006711211.1 | 3 - 4 | 399 | 121 | ||
| GenBank mRNA | AF288394.1 | - | 3 - 4 | 425 | 121 | |
| AK098196.1 | - | 3 - 4 | 348 | 121 | ||
| BC022030.1 | - | 3 - 4 | 395 | 4824025 | 121 | |
| BI821111.1 | - | 3 - 4 | 375 | 121 | ||
| BP326115.1 | - | 3 - 4 | 375 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

TSEN15
Genomic Map