Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
TC01001700.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
ENST00000367182.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
ENST00000391947.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
ENST00000454264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
ENST00000463049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
ENST00000543518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
NM_001204171.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026417.hg.1 |
NM_002393.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
TC01001700.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
ENST00000367182.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
ENST00000454264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
ENST00000463049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
ENST00000543518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
NM_001204171.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026412.hg.1 |
NM_002393.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000367182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000454264.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000463049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000614459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000621032.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_001204171.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_001278517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_002393.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000367182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000454264.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000463049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000614459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
ENST00000621032.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_001204171.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_001278517.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
NM_002393.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240289.hg.1 |
TC0100018339.hg.1 |
Clariom D TC |