Assay Details
| Gene Symbol: TMEM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033504.2 | NP_277039.1 | 4 - 5 | 571 | 89 | |
| XM_005270408.3 | XP_005270465.1 | 4 - 5 | 654 | 89 | ||
| GenBank mRNA | AY027543.1 | - | 4 - 5 | 508 | 89 | |
| AY359883.1 | - | 3 - 4 | 397 | 89 | ||
| BC001418.3 | - | 4 - 5 | 571 | 3140820 | 89 | |
| BC013953.2 | - | 4 - 5 | 571 | 2987806 | 89 | |
| BC023260.1 | - | 3 - 4 | 449 | 4213210 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002477.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01038281.hg.1 | TC01002477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038281.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038281.hg.1 | ENST00000373463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038281.hg.1 | ENST00000475208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038281.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038283.hg.1 | TC01002477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038283.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038283.hg.1 | ENST00000373463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038283.hg.1 | ENST00000475208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038283.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038284.hg.1 | TC01002477.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038284.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038284.hg.1 | ENST00000373463.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038284.hg.1 | ENST00000475208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01038284.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201417.hg.1 | TC0100013634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201419.hg.1 | TC0100013634.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | ENST00000482771.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013634.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000329151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000373463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000463914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000474144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | ENST00000475208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | NM_033504.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100201420.hg.1 | TC0100013634.hg.1 | Clariom D TC |

TMEM54
Genomic Map