Assay Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015284.3 | NP_056099.3 | 34 - 35 | 5003 | 61 | |
| XM_005270686.3 | XP_005270743.1 | 36 - 37 | 5235 | 61 | ||
| XM_006710501.3 | XP_006710564.1 | 35 - 36 | 5122 | 61 | ||
| XM_011541106.2 | XP_011539408.1 | 35 - 36 | 5273 | 61 | ||
| XM_011541107.2 | XP_011539409.1 | 32 - 33 | 4770 | 61 | ||
| XM_017000819.1 | XP_016856308.1 | 36 - 37 | 5235 | 61 | ||
| XM_017000820.1 | XP_016856309.1 | 35 - 36 | 5064 | 61 | ||
| XM_017000821.1 | XP_016856310.1 | 32 - 33 | 4756 | 61 | ||
| GenBank mRNA | AB007936.2 | - | 20 - 21 | 2855 | 61 | |
| BC151232.1 | - | 20 - 21 | 2855 | 8860124 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000547.hg.1
Set Membership: |
|

SZT2
Genomic Map