Assay Details
| Gene Symbol: C6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109938.2 | NP_001103408.1 | 5 - 6 | 1029 | 68 | |
| NM_001161376.1 | NP_001154848.1 | 5 - 6 | 1572 | 68 | ||
| NM_145029.3 | NP_659466.2 | 4 - 5 | 627 | 68 | ||
| XM_011514384.1 | XP_011512686.1 | 6 - 7 | 1121 | 68 | ||
| GenBank mRNA | BC016167.1 | - | 5 - 6 | 1180 | 3509511 | 68 |
| BC064397.1 | - | 5 - 6 | 1190 | 68 | ||
| BM695485.1 | - | 4 - 5 | 502 | 68 | ||
| BQ022831.1 | - | 1 - 2 | 419 | 68 | ||
More Information
Set Membership: |
|

C6orf136
Genomic Map