Assay Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318316.1 | NP_001305245.1 | 4 - 5 | 382 | 80 | |
| NM_172002.4 | NP_741999.3 | 4 - 5 | 628 | 80 | ||
| NR_134560.1 | 4 - 5 | 573 | 80 | |||
| XM_005261359.3 | XP_005261416.1 | 4 - 5 | 629 | 80 | ||
| XM_017028619.1 | XP_016884108.1 | 4 - 5 | 599 | 80 | ||
| XM_017028620.1 | XP_016884109.1 | 4 - 5 | 432 | 80 | ||
| GenBank mRNA | AY191719.1 | - | 4 - 5 | 578 | 80 | |
| BC000004.1 | - | 4 - 5 | 637 | 80 | ||
| BC065569.1 | - | 4 - 5 | 601 | 6171959 | 80 | |
| CR456462.1 | - | 4 - 5 | 596 | 80 | ||
| HY057929.1 | - | 4 - 5 | 382 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000186.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22003419.hg.1 | ENST00000216027.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003419.hg.1 | ENST00000398941.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003419.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003419.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003419.hg.1 | TC22000186.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000485599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007001.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | TC2200007001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | NM_172002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000216027.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000398941.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000420442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000450178.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000483861.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | ENST00000495977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148763.hg.1 | NM_172002.4 | Refseq |

HSCB
Genomic Map