| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026713.hg.1 |
ENST00000299855.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026713.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026713.hg.1 |
TC11002235.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026714.hg.1 |
ENST00000299855.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026714.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026714.hg.1 |
TC11002235.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026711.hg.1 |
ENST00000299855.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026711.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026711.hg.1 |
TC11002235.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026712.hg.1 |
ENST00000299855.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026712.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026712.hg.1 |
TC11002235.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100193277.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193277.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193277.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193277.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193277.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100193278.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193278.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193278.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193278.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193278.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100193279.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193279.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193279.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193279.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193279.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000434103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
ENST00000524478.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012133.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193276.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193276.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193276.hg.1 |
ENST00000299855.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193276.hg.1 |
NM_002422.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193276.hg.1 |
TC1100012133.hg.1 |
Clariom D TC |