Assay Details
| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291746.1 | NP_001278675.1 | 7 - 8 | 1198 | 124 | |
| NM_002908.3 | NP_002899.1 | 7 - 8 | 1198 | 124 | ||
| XM_011533010.2 | XP_011531312.1 | 5 - 6 | 685 | 124 | ||
| XM_017004627.1 | XP_016860116.1 | 7 - 8 | 1119 | 124 | ||
| GenBank mRNA | AK303926.1 | - | 7 - 8 | 1119 | 124 | |
| BC117191.1 | - | 7 - 8 | 1037 | 40125742 | 124 | |
| BC143885.1 | - | 7 - 8 | 1039 | 9052401 | 124 | |
| X75042.1 | - | 7 - 8 | 1029 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000348.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
inflammatory response
I-kappaB kinase/NF-kappaB signaling
negative regulation of gene expression
negative regulation of interferon-beta production
response to cytokine
NIK/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of transcription from RNA polymerase II promoter
negative regulation of neuron death
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
inflammatory response
I-kappaB kinase/NF-kappaB signaling
negative regulation of gene expression
negative regulation of interferon-beta production
response to cytokine
NIK/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of transcription from RNA polymerase II promoter
negative regulation of neuron death
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02005178.hg.1 | TC02000348.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005178.hg.1 | ENST00000295025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005178.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005180.hg.1 | TC02000348.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005180.hg.1 | ENST00000295025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005180.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005180.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000589496.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | ENST00000589496.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005179.hg.1 | TC02000348.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005179.hg.1 | ENST00000295025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005179.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005179.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000589496.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | ENST00000589496.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016452.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221897.hg.1 | TC0200016452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | ENST00000394479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | NM_001291746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221899.hg.1 | TC0200016452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200221898.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221898.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221898.hg.1 | ENST00000295025.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200221898.hg.1 | NM_002908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200221898.hg.1 | TC0200016452.hg.1 | Clariom D TC |

REL
Genomic Map