Assay Details
| Gene Symbol: ARG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000045.3 | NP_000036.2 | 2 - 3 | 211 | 63 | |
| XM_011535801.2 | XP_011534103.1 | 2 - 3 | 256 | 63 | ||
| GenBank mRNA | BC005321.1 | - | 2 - 3 | 189 | 3934479 | 63 |
| BC020653.1 | - | 2 - 3 | 137 | 4712189 | 63 | |
| BG542163.1 | - | 213 | 63 | |||
| BT006741.1 | - | 2 - 3 | 132 | 63 | ||
| M14502.1 | - | 2 - 3 | 188 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000983.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
urea cycle
liver development
positive regulation of endothelial cell proliferation
arginine catabolic process
aging
response to herbicide
response to manganese ion
response to zinc ion
response to selenium ion
regulation of L-arginine import
response to amine
arginine catabolic process to ornithine
lung development
collagen biosynthetic process
response to vitamin A
response to vitamin E
response to drug
response to amino acid
response to cadmium ion
response to axon injury
response to methylmercury
mammary gland involution
maternal process involved in female pregnancy
protein homotrimerization
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
cellular response to interleukin-4
cellular response to glucagon stimulus
cellular response to dexamethasone stimulus
cellular response to transforming growth factor beta stimulus
arginase activity
manganese ion binding
urea cycle
liver development
positive regulation of endothelial cell proliferation
arginine catabolic process
aging
response to herbicide
response to manganese ion
response to zinc ion
response to selenium ion
regulation of L-arginine import
response to amine
arginine catabolic process to ornithine
lung development
collagen biosynthetic process
response to vitamin A
response to vitamin E
response to drug
response to amino acid
response to cadmium ion
response to axon injury
response to methylmercury
mammary gland involution
maternal process involved in female pregnancy
protein homotrimerization
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
cellular response to interleukin-4
cellular response to glucagon stimulus
cellular response to dexamethasone stimulus
cellular response to transforming growth factor beta stimulus
arginase activity
manganese ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | ENST00000368087.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | ENST00000476845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | ENST00000489091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06011585.hg.1 | TC06000983.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600164835.hg.1 | TC0600009455.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000275196.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000356962.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000368087.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000469293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000484820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | ENST00000498260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_000045.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009455.hg.1 | NM_001244438.1 | Refseq |

ARG1
Genomic Map