Assay Details
| Gene Symbol: KIF26B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018012.3 | NP_060482.2 | 10 - 11 | 2704 | 64 | |
| GenBank mRNA | AK128806.1 | - | 1 - 2 | 197 | 64 | |
| AK297303.1 | - | 10 - 11 | 2700 | 64 | ||
| AY923834.1 | - | 10 - 11 | 2705 | 64 | ||
| BC035896.1 | - | 7 - 8 | 1225 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001996.hg
Additional Information:
This assay is in the 5'UTR region of the AK128806.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
axon guidance
protein localization
positive regulation of cell-cell adhesion
establishment of cell polarity
cytoskeleton-dependent intracellular transport
ureteric bud invasion
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
microtubule-based movement
axon guidance
protein localization
positive regulation of cell-cell adhesion
establishment of cell polarity
cytoskeleton-dependent intracellular transport
ureteric bud invasion
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030921.hg.1 | TC01001996.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030921.hg.1 | ENST00000366518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030921.hg.1 | ENST00000407071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030921.hg.1 | ENST00000413001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030921.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190150.hg.1 | TC0100012247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000366518.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000407071.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000479506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | ENST00000483253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012247.hg.1 | NM_018012.4 | Refseq |

KIF26B
Genomic Map