| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000340077.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000346874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000350537.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000374773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000374776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000374788.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000374789.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000374790.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000466092.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000544292.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000545260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
ENST00000545693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184785.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184786.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184787.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184789.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184793.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_001184794.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
NM_019619.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014767.hg.1 |
TC10001170.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000340077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000346874.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000350537.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374776.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374788.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374790.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000466092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000544292.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000545260.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000545693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184787.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_019619.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000340077.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000346874.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000350537.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374773.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374776.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374788.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000374790.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000466092.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000544292.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000545260.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
ENST00000545693.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184785.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184786.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184787.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184793.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_001184794.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
NM_019619.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000170141.hg.1 |
TC1000010292.hg.1 |
Clariom D TC |