Assay Details
| Gene Symbol: PARD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184793.1 | NP_001171722.1 | 18 - 19 | 2840 | 92 | |
| GenBank mRNA | AK027735.1 | - | 16 - 17 | 2137 | 92 | |
More Information
Set Membership: |
|

PARD3
Genomic Map