Assay Details
| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024105.3 | NP_077010.1 | 8 - 9 | 1437 | 110 | |
| XM_011530371.2 | XP_011528673.1 | 8 - 9 | 1326 | 110 | ||
| XM_017028936.1 | XP_016884425.1 | 8 - 9 | 1448 | 110 | ||
| XM_017028937.1 | XP_016884426.1 | 8 - 9 | 1448 | 110 | ||
| GenBank mRNA | AF258571.1 | - | 4 - 5 | 566 | 110 | |
| AF318343.1 | - | 8 - 9 | 1389 | 110 | ||
| AJ290427.1 | - | 8 - 9 | 1310 | 110 | ||
| AJ303120.1 | - | 8 - 9 | 1310 | 110 | ||
| BC001729.1 | - | 8 - 9 | 1284 | 3353085 | 110 | |
| BC098562.1 | - | 8 - 9 | 1330 | 30560020 | 110 | |
| CR456369.1 | - | 7 - 8 | 1226 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000904.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015103.hg.1 | ENST00000330817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015103.hg.1 | ENST00000332276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015103.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015103.hg.1 | TC22000904.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015104.hg.1 | ENST00000330817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015104.hg.1 | ENST00000332276.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015104.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015104.hg.1 | TC22000904.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | TC2200009128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165962.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009128.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | TC2200009128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | NM_024105.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000330817.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000486602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | ENST00000492791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165961.hg.1 | NM_024105.3 | Refseq |

ALG12
Genomic Map