Assay Details
| Gene Symbol: GALNT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017423.2 | NP_059119.2 | 10 - 11 | 1793 | 106 | |
| XM_005263062.1 | XP_005263119.1 | 10 - 11 | 1793 | 106 | ||
| XM_011532025.2 | XP_011530327.1 | 10 - 11 | 2540 | 106 | ||
| XM_017008292.1 | XP_016863781.1 | 13 - 14 | 2713 | 106 | ||
| XM_017008293.1 | XP_016863782.1 | 13 - 14 | 4502 | 106 | ||
| GenBank mRNA | AJ002744.1 | - | 10 - 11 | 1742 | 106 | |
| AJ505958.1 | - | 2 - 3 | 237 | 106 | ||
| AK128437.1 | - | 1 - 2 | 1879 | 106 | ||
| AK295509.1 | - | 8 - 9 | 1099 | 106 | ||
| BC035303.1 | - | 9 - 10 | 1224 | 106 | ||
| BC046129.1 | - | 10 - 11 | 1771 | 4442080 | 106 | |
| BC047468.1 | - | 10 - 11 | 1768 | 5271749 | 106 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000853.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04011732.hg.1 | TC04000853.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011732.hg.1 | ENST00000265000.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011732.hg.1 | ENST00000458613.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011732.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400163961.hg.1 | TC0400009322.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000265000.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000502407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000503213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000505308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000506317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000512285.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | ENST00000515862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009322.hg.1 | NM_017423.2 | Refseq |

GALNT7
Genomic Map