Assay Details
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317837.1 | NP_001304766.1 | 4 - 5 | 457 | 102 | |
NM_001317838.1 | NP_001304767.1 | 4 - 5 | 454 | 102 | ||
NM_001317839.1 | NP_001304768.1 | 4 - 5 | 332 | 102 | ||
NM_152399.3 | NP_689612.3 | 4 - 5 | 613 | 102 | ||
XM_005262737.2 | XP_005262794.1 | 4 - 5 | 421 | 102 | ||
XM_017007725.1 | XP_016863214.1 | 4 - 5 | 327 | 102 | ||
GenBank mRNA | AK055396.1 | - | 4 - 5 | 334 | 102 | |
AL552582.3 | - | 4 - 5 | 484 | 102 | ||
BC047657.1 | - | 4 - 5 | 613 | 5264721 | 102 | |
DA176165.1 | - | 4 - 5 | 418 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001515.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK055396.1,BC047657.1,NM_001317837.1,NM_001317838.1,NM_001317839.1,NM_152399.3,XM_005262737.2,XM_017007725.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021515.hg.1 | TC04001515.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021515.hg.1 | ENST00000337677.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021515.hg.1 | ENST00000394394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021515.hg.1 | ENST00000394396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021515.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021513.hg.1 | TC04001515.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021513.hg.1 | ENST00000337677.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021513.hg.1 | ENST00000394394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021513.hg.1 | ENST00000394396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021513.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000506551.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400011746.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180797.hg.1 | TC0400011746.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000337677.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000394394.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000394396.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000461198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000505198.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000513254.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | ENST00000514885.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | NM_152399.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400180799.hg.1 | TC0400011746.hg.1 | Clariom D TC |
