Assay Details
| Gene Symbol: LOC107984562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020896.1 | XP_016876385.1 | 1 - 2 | 665 | 86 | |
| XM_017020897.1 | XP_016876386.1 | 2 - 3 | 250 | 86 | ||
| XM_017020898.1 | XP_016876387.1 | 2 - 3 | 253 | 86 | ||
More Information
Set Membership: |
|

LOC107984562
Genomic Map