| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004322.hg.1 |
ENST00000299413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004322.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004322.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004324.hg.1 |
ENST00000299413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004324.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004324.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004320.hg.1 |
ENST00000299413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004320.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004320.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004326.hg.1 |
ENST00000299413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004326.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004326.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004329.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004329.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004327.hg.1 |
ENST00000299413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004327.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11004327.hg.1 |
TC11000339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000570295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151431.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151434.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151434.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151434.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151434.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151434.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151432.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151436.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151436.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151436.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151436.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151436.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151433.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151433.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151433.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151433.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151433.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100151435.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151435.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151435.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151435.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151435.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000570295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |