Assay Details
| Gene Symbol: MAGOHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135120.1 | 3 - 4 | 341 | 152 | ||
| NR_135121.1 | 3 - 4 | 239 | 152 | |||
| GenBank mRNA | BG539870.1 | - | 3 - 4 | 222 | 152 | |
| CD357202.1 | - | 3 - 4 | 344 | 152 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000320756.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000398874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000537852.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000539554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000540074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000543929.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000544850.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000545236.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000546173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | ENST00000625272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_001300739.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009911.hg.1 | NM_018048.4 | Refseq |

MAGOHB
Genomic Map