Assay Details
| Gene Symbol: TMBIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282606.1 | NP_001269535.1 | 2 - 3 | 356 | 78 | |
| GenBank mRNA | AF182414.1 | - | 2 - 3 | 243 | 78 | |
More Information
Set Membership: |
|

TMBIM4
Genomic Map