Assay Details
| Gene Symbol: TLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006289.3 | NP_006280.3 | 50 - 51 | 7119 | 65 | |
| GenBank mRNA | AB028950.2 | - | 50 - 51 | 7094 | 65 | |
| AF078828.1 | - | 50 - 51 | 6891 | 65 | ||
| AF177198.1 | - | 50 - 51 | 6855 | 65 | ||
| BC042923.1 | - | 50 - 51 | 6905 | 5576203 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001059.hg.1
Set Membership: |
|

TLN1
Genomic Map