Assay Details
| Gene Symbol: CLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001831.3 | NP_001822.3 | 5 - 6 | 1015 | 62 | |
| NR_038335.1 | 5 - 6 | 1150 | 62 | |||
| NR_045494.1 | 5 - 6 | 1009 | 62 | |||
| XM_006716284.2 | XP_006716347.1 | 5 - 6 | 2921 | 62 | ||
| GenBank mRNA | AK093399.1 | - | 6 - 7 | 765 | 62 | |
| AK301324.1 | - | 5 - 6 | 619 | 62 | ||
| AK313870.1 | - | 5 - 6 | 904 | 62 | ||
| AY513288.1 | - | 5 - 6 | 894 | 62 | ||
| BC010514.1 | - | 5 - 6 | 897 | 4150452 | 62 | |
| BC013221.1 | - | 4 - 5 | 830 | 62 | ||
| BC019588.2 | - | 4 - 5 | 865 | 4915444 | 62 | |
| BC029972.1 | - | 5 - 6 | 897 | 62 | ||
| BX459023.2 | - | 5 - 6 | 888 | 62 | ||
| BX648414.1 | - | 4 - 5 | 1615 | 62 | ||
| EB385800.1 | - | 3 - 4 | 662 | 62 | ||
| J02908.1 | - | 3 - 4 | 747 | 62 | ||
| M25915.1 | - | 5 - 6 | 1024 | 62 | ||
| M64722.1 | - | 5 - 6 | 881 | 62 | ||
| M74816.1 | - | 3 - 4 | 651 | 62 | ||
| X14723.1 | - | 4 - 5 | 876 | 62 | ||
More Information
Set Membership: |
|

CLU
Genomic Map