Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243736.1 | NP_001230665.1 | 5 - 6 | 466 | 108 | |
| NM_021970.3 | NP_068805.1 | 5 - 6 | 487 | 108 | ||
| NR_024170.1 | 4 - 5 | 441 | 108 | |||
| GenBank mRNA | AF201947.1 | - | 5 - 6 | 397 | 108 | |
| AK223311.1 | - | 5 - 6 | 378 | 108 | ||
| AK311755.1 | - | 5 - 6 | 348 | 108 | ||
| AL544981.3 | - | 5 - 6 | 296 | 108 | ||
| BC026245.1 | - | 5 - 6 | 380 | 4808855 | 108 | |
| BC110402.1 | - | 4 - 5 | 307 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001416.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
cell cycle arrest
macroautophagy
positive regulation of TOR signaling
cellular protein localization
positive regulation of GTPase activity
cellular response to amino acid stimulus
guanyl-nucleotide exchange factor activity
protein binding
protein complex scaffold
MAPK cascade
cell cycle arrest
macroautophagy
positive regulation of TOR signaling
cellular protein localization
positive regulation of GTPase activity
cellular response to amino acid stimulus
guanyl-nucleotide exchange factor activity
protein binding
protein complex scaffold
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | TC04001416.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | ENST00000499666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04019772.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400178160.hg.1 | TC0400011421.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000226522.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000499666.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | ENST00000515100.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_001243736.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NM_021970.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011421.hg.1 | NR_024170.1 | Refseq |

LAMTOR3
Genomic Map