Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300993.2 | NP_001287922.1 | 4 - 5 | 450 | 88 | |
| NM_001321991.1 | NP_001308920.1 | 3 - 4 | 299 | 88 | ||
| NM_001321992.1 | NP_001308921.1 | 2 - 3 | 630 | 88 | ||
| NM_001321993.1 | NP_001308922.1 | 3 - 4 | 714 | 88 | ||
| NM_001321994.1 | NP_001308923.1 | 2 - 3 | 630 | 88 | ||
| NM_144694.3 | NP_653295.1 | 3 - 4 | 714 | 88 | ||
| XM_011526544.2 | XP_011524846.1 | 3 - 4 | 297 | 88 | ||
| XM_011526545.2 | XP_011524847.1 | 2 - 3 | 201 | 88 | ||
| GenBank mRNA | AK055353.1 | - | 3 - 4 | 692 | 88 | |
| AK297561.1 | - | 2 - 3 | 332 | 88 | ||
| AK308252.1 | - | 4 - 5 | 450 | 88 | ||
| BC130425.1 | - | 3 - 4 | 597 | 40146456 | 88 | |
| BC130433.1 | - | 3 - 4 | 597 | 40146464 | 88 | |
| BP312352.1 | - | 3 - 4 | 299 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000516.hg
Additional Information:
This assay is in the 5'UTR region of the AK297561.1,NM_001321993.1,NM_001321994.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19006860.hg.1 | ENST00000330173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006860.hg.1 | ENST00000388801.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006860.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19006860.hg.1 | TC19000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | ENST00000591976.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | TC1900007975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900159638.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000330173.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000586475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000587363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000588644.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000589725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000590664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591380.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | ENST00000591976.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_001300993.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007975.hg.1 | NM_144694.3 | Refseq |

ZNF570
Genomic Map