| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015853.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015853.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050160.hg.1 |
TC01003256.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050160.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050160.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050162.hg.1 |
TC01003256.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050162.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219054.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219054.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219054.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219054.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219054.hg.1 |
TC0100015853.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015853.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015853.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015853.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219053.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219053.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219053.hg.1 |
ENST00000368748.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219053.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219053.hg.1 |
TC0100015853.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219058.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219058.hg.1 |
NM_001014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219058.hg.1 |
TC0100015853.hg.1 |
Clariom D TC |