Assay Details
| Gene Symbol: SYDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300910.1 | NP_001287839.1 | 3 - 4 | 947 | 95 | |
| NM_033025.5 | NP_149014.3 | 3 - 4 | 1148 | 95 | ||
| GenBank mRNA | AK123686.1 | - | 3 - 4 | 1109 | 95 | |
| AK315453.1 | - | 3 - 4 | 906 | 95 | ||
| BC029926.1 | - | 3 - 4 | 889 | 5106728 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000269.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19003838.hg.1 | ENST00000342784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003838.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19003838.hg.1 | TC19000269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000597977.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007208.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | TC1900007208.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152074.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | NM_033025.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | TC1900007208.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000342784.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000600252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000600440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | ENST00000602203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | NM_001300910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900152075.hg.1 | NM_033025.5 | Refseq |

SYDE1
Genomic Map