Assay Details
| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324142.1 | NP_001311071.1 | 2 - 3 | 432 | 114 | |
| NM_001324148.1 | NP_001311077.1 | 2 - 3 | 522 | 114 | ||
| NM_001324156.1 | NP_001311085.1 | 1 - 2 | 256 | 114 | ||
| GenBank mRNA | AJ010017.1 | - | 1 - 2 | 135 | 114 | |
| AJ010022.1 | - | 1 - 2 | 135 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ010017.1, AJ010022.1, NM_001324142.1, NM_001324148.1, NM_001324156.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000313116.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000377065.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000432977.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | ENST00000465311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_007130.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009569.hg.1 | NM_153380.3 | Refseq |

ZNF41
Genomic Map