Assay Details
Gene Symbol: PRDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012406.3 | NP_036538.3 | 7 - 8 | 1833 | 110 | |
XM_005268593.1 | XP_005268650.1 | 7 - 8 | 1852 | 110 | ||
GenBank mRNA | AF144757.2 | - | 6 - 7 | 1518 | 110 | |
AK056378.1 | - | 7 - 8 | 1700 | 110 | ||
AL133083.1 | - | 4 - 5 | 1168 | 110 | ||
BC035581.1 | - | 6 - 7 | 1634 | 5587364 | 110 | |
BC068562.1 | - | 7 - 8 | 1534 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001928.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
cell proliferation
negative regulation of cell growth
cellular response to nutrient
histone H4-R3 methylation
cellular response to growth factor stimulus
DNA binding
death receptor binding
protein binding
methyltransferase activity
zinc ion binding
histone methyltransferase binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
cell proliferation
negative regulation of cell growth
cellular response to nutrient
histone H4-R3 methylation
cellular response to growth factor stimulus
DNA binding
death receptor binding
protein binding
methyltransferase activity
zinc ion binding
histone methyltransferase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12025111.hg.1 | ENST00000228437.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025111.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025111.hg.1 | TC12001928.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025112.hg.1 | ENST00000228437.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12025112.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12025112.hg.1 | TC12001928.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000547268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011820.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191589.hg.1 | TC1200011820.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000228437.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000549416.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000550376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | ENST00000550659.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | NM_012406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200191587.hg.1 | TC1200011820.hg.1 | Clariom D TC |
