Assay Details
| Gene Symbol: LRRC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098519.1 | NP_001091989.1 | 8 - 9 | 1517 | 96 | |
| NM_152759.4 | NP_689972.3 | 8 - 9 | 1406 | 96 | ||
| XM_017019124.1 | XP_016874613.1 | 8 - 9 | 1525 | 96 | ||
| GenBank mRNA | AK124107.1 | - | 10 - 11 | 1786 | 96 | |
| BC029245.2 | - | 7 - 8 | 1341 | 5169080 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000970.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | ENST00000289014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | ENST00000425921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012699.hg.1 | TC12000970.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | ENST00000289014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | ENST00000425921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012700.hg.1 | TC12000970.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | ENST00000289014.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | ENST00000425921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012698.hg.1 | TC12000970.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168208.hg.1 | TC1200009198.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168209.hg.1 | TC1200009198.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537729.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000537733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000538800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009198.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | ENST00000339777.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | ENST00000541498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | NM_001098519.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | NM_152759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200168207.hg.1 | TC1200009198.hg.1 | Clariom D TC |

LRRC43
Genomic Map