Assay Details
Gene Symbol: C18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145055.3 | NP_659492.1 | 2 - 3 | 1094 | 84 | |
XM_011525822.1 | XP_011524124.1 | 2 - 3 | 945 | 84 | ||
GenBank mRNA | BC016149.1 | - | 1 - 2 | 767 | 4040087 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000167.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000415357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000415357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001953.hg.1 | ENST00000282059.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001953.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001953.hg.1 | TC18000167.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000415357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000415357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000434418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | ENST00000632331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015197.hg.1 | NR_126396.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149533.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149533.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149533.hg.1 | TC1800007218.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800149533.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149533.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | NM_145055.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | TC1800007218.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | ENST00000588730.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | ENST00000615052.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800149532.hg.1 | NM_145055.3 | Refseq |
