Assay Details
| Gene Symbol: C18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145055.3 | NP_659492.1 | 3 - 4 | 1280 | 64 | |
| XM_011525822.1 | XP_011524124.1 | 3 - 4 | 1131 | 64 | ||
| GenBank mRNA | BC016149.1 | - | 2 - 3 | 953 | 4040087 | 64 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |

C18orf25
Genomic Map