Assay Details
| Gene Symbol: C18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008239.2 | NP_001008240.1 | 3 - 4 | 1216 | 102 | |
| NM_145055.3 | NP_659492.1 | 4 - 5 | 1399 | 102 | ||
| XM_011525822.1 | XP_011524124.1 | 4 - 5 | 1250 | 102 | ||
| XM_017025555.1 | XP_016881044.1 | 3 - 4 | 1063 | 102 | ||
| GenBank mRNA | AK289738.1 | - | 3 - 4 | 1100 | 102 | |
| AK292981.1 | - | 3 - 4 | 1204 | 102 | ||
| AL713661.1 | - | 2 - 3 | 396 | 102 | ||
| AL832027.2 | - | 2 - 3 | 744 | 102 | ||
| BC016149.1 | - | 3 - 4 | 1072 | 4040087 | 102 | |
| BC084579.1 | - | 3 - 4 | 1183 | 6141272 | 102 | |
| BX640683.1 | - | 2 - 3 | 897 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000167.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18001957.hg.1 | ENST00000282059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001957.hg.1 | ENST00000321319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001957.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001957.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001957.hg.1 | TC18000167.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001958.hg.1 | ENST00000282059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001958.hg.1 | ENST00000321319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001958.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001958.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001958.hg.1 | TC18000167.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | TC1800007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149537.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | TC1800007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149536.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | TC1800007218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800149535.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000587591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000588730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615052.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000615553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619230.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | ENST00000619301.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_001008239.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007218.hg.1 | NM_145055.3 | Refseq |

C18orf25
Genomic Map