Assay Details
| Gene Symbol: PNLIPRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017016339.1 | XP_016871828.1 | 2 - 3 | 218 | 91 | |
| GenBank mRNA | AK308508.1 | - | 3 - 4 | 244 | 91 | |
| AK308521.1 | - | 3 - 4 | 244 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017016339.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000579578.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000586762.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000588823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000591655.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | ENST00000611850.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NM_005396.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009007.hg.1 | NR_103727.1 | Refseq |

PNLIPRP2
Genomic Map