Assay Details
| Gene Symbol: ENTPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017021814.1 | XP_016877303.1 | 1 - 2 | 121 | 72 | |
| XM_017021816.1 | XP_016877305.1 | 1 - 2 | 121 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017021814.1, XM_017021816.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
cell growth
positive regulation of glycolytic process
ATP metabolic process
'de novo' posttranslational protein folding
guanosine-diphosphatase activity
protein binding
uridine-diphosphatase activity
protein N-linked glycosylation
cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
cell growth
positive regulation of glycolytic process
ATP metabolic process
'de novo' posttranslational protein folding
guanosine-diphosphatase activity
protein binding
uridine-diphosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000334696.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000553284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000554664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000555829.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556108.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000556242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557325.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | ENST00000557681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009682.hg.1 | NM_001249.3 | Refseq |

ENTPD5
Genomic Map