| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013389.hg.1 |
TC06001126.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013389.hg.1 |
ENST00000252631.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013389.hg.1 |
ENST00000392185.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013389.hg.1 |
ENST00000539592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013389.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013388.hg.1 |
TC06001126.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013388.hg.1 |
ENST00000252631.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013388.hg.1 |
ENST00000392185.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013388.hg.1 |
ENST00000539592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06013388.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014196.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
ENST00000392185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
ENST00000611124.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
NM_016224.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600199781.hg.1 |
TC0600014196.hg.1 |
Clariom D TC |