Assay Details
| Gene Symbol: SHANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012309.4 | NP_036441.2 | 24 - 25 | 5087 | 80 | |
| NM_133266.4 | NP_573573.2 | 10 - 11 | 3284 | 80 | ||
| XM_005277932.3 | XP_005277989.1 | 15 - 16 | 4092 | 80 | ||
| XM_017017387.1 | XP_016872876.1 | 25 - 26 | 5192 | 80 | ||
| XM_017017388.1 | XP_016872877.1 | 25 - 26 | 5214 | 80 | ||
| XM_017017389.1 | XP_016872878.1 | 25 - 26 | 5360 | 80 | ||
| XM_017017390.1 | XP_016872879.1 | 12 - 13 | 6325 | 80 | ||
| GenBank mRNA | AB028945.1 | - | 7 - 8 | 2821 | 80 | |
| AB208025.1 | - | 24 - 25 | 5053 | 80 | ||
| AB208026.1 | - | 15 - 16 | 3875 | 80 | ||
| AB208027.1 | - | 10 - 11 | 3262 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

SHANK2
Genomic Map