Assay Details
| Gene Symbol: PRPF38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018061.2 | NP_060531.2 | 1 - 2 | 560 | 60 | |
| XM_011541687.2 | XP_011539989.1 | 1 - 2 | 578 | 60 | ||
| GenBank mRNA | AK001192.1 | - | 1 - 2 | 354 | 60 | |
| BC007757.2 | - | 1 - 2 | 534 | 60 | ||
| BC009453.1 | - | 1 - 2 | 530 | 60 | ||
| BC016296.1 | - | 1 - 2 | 530 | 60 | ||
| BC024275.1 | - | 1 - 2 | 534 | 60 | ||
| BC040127.1 | - | 1 - 2 | 547 | 60 | ||
| BC053838.1 | - | 1 - 2 | 546 | 60 | ||
| BC107801.1 | - | 1 - 2 | 287 | 60 | ||
| BC130346.1 | - | 1 - 2 | 306 | 40146377 | 60 | |
| BC132963.1 | - | 1 - 2 | 306 | 40146985 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000932.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | TC01000932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | ENST00000370022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | ENST00000370025.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | ENST00000467302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014628.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | TC01000932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | ENST00000370022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | ENST00000370025.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | ENST00000467302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014631.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | TC01000932.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | ENST00000370022.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | ENST00000370025.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | ENST00000467302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014629.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166425.hg.1 | TC0100009328.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166423.hg.1 | TC0100009328.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | ENST00000485810.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009328.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166424.hg.1 | TC0100009328.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370022.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000370025.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | ENST00000467302.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | NM_018061.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | NR_037185.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166427.hg.1 | TC0100009328.hg.1 | Clariom D TC |

PRPF38B
Genomic Map